Disposable columns prepacked with Sepharose™, GST GraviTrap™
Fournisseur: Cytiva
28-9523-60
28-9523-60EA 672 EUR InStock
28-9523-60
Disposable columns prepacked with Sepharose™, GST GraviTrap™
Colonnes pour chromatographie
GST GraviTrap contains 10 disposable columns prepacked with 2 ml of Glutathione Sepharose 4B, sufficient for purification of up to 20 mg of fusion protein.
  • Convenient, prepacked gravity-flow columns
  • Allows direct purification of both clarified and unclarified cell lysates
  • Yields up to 20 mg of pure proteins per column
GST GraviTrap contains 10 disposable columns prepacked with 2 mL of Glutathione Sepharose 4B, sufficient for purification of up to 20 mg of fusion protein. GST-tagged proteins are recovered under mild elution conditions (10 mM glutathione), which preserves functionality of the proteins.
The binding capacity of prepacked GST GraviTrap columns, which can be greater than 10 mg protein/mL medium, varies with the nature of the tagged protein and the exact binding conditions used. For increased convenience, GST Buffer Kit contains stock solutions of the buffers needed for purification of GST-tagged proteins.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité