Affinity chromatography columns, rProtein A Protein G GraviTrap™
Fournisseur: Cytiva
28-9852-55
28-9852-55EA 951 EUR InStock
28-9852-55
Affinity chromatography columns, rProtein A Protein G GraviTrap™
Colonnes pour chromatographie
Protein G GraviTrap includes ten columns with 1 ml of resin for general purpose purification of polyclonal and monoclonal antibodies from most species.
  • Broad binding specificity – binds a broad range of IgG species and subclasses from most species including rat
  • High binding capacity – approximately 20 mg human IgG/ml resin
  • Simple – GraviTrap prepacked gravity flow columns enable simple one-step purification of proteins in 30 min without the need for equipment
Protein G GraviTrap columns are prepacked with Protein G Sepharose 4 Fast Flow resin. These gravity flow columns are designed for fast and efficient manual purification of monoclonal and polyclonal antibodies. Each of these protein G columns is reusable up to five times. Prepacked gravity flow columns provide convenient, fast, and reliable results.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité