Affinity chromatography columns, Protein G GraviTrap™
Fournisseur: Cytiva
28-9852-56
28-9852-56EA 980 EUR InStock
28-9852-56
Affinity chromatography columns, Protein G GraviTrap™
Colonnes pour chromatographie
rProtein A Protein G GraviTrap contains ten columns with 1 ml 50% rProtein A Sepharose Fast Flow and 50% Protein G Sepharose 4 Fast Flow resins.
  • For general-purpose purification of monoclonal and polyclonal antibodies
  • Suitable when the affinity for protein A and protein G is not known
  • GraviTrap prepacked gravity flow columns enable simple one-step purification of proteins in 30 min without the need for equipment
  • High purity: >95% in one step
  • High protein recovery (typically 70% to 80%) due to high binding capacity of ~35 mg IgG/ml resin
This column can be used for manual purification of polyclonal and monoclonal antibodies from cell culture supernatant and biological fluids. It is particularly suitable when the affinity for protein A and protein G is not known. The gravity flow column relies on gravity and is reusable up to five times, so no additional instruments are needed for this protocol.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité