Hydrophobic interaction chromatography columns, HiScreen™ Capto™ Phenyl
Fournisseur: Cytiva
28-9924-72
28-9924-72EA 171 EUR InStock
28-9924-72
Hydrophobic interaction chromatography columns, HiScreen™ Capto™ Phenyl
Colonnes pour chromatographie
Capto™ Phenyl (high sub) and Capto™ Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
  • Excellent for method optimization and parameter screening due to the 10 cm bed height
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume for fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same resin using the same flow velocity
Capto™ Phenyl (high sub) is an excellent choice for initial and intermediate step purifications requiring a chromatography resin with resin to high hydrophobicity. Performance and selectivity are similar to that of Phenyl Sepharose™ 6 Fast Flow (high sub). Higher flow rates can be used with Capto™ Phenyl (high sub) during equilibration, wash and regeneration.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité