Affinity chromatography columns, His SpinTrap™ TALON®
Fournisseur: Cytiva
29-0005-93
29-0005-93EA 416 EUR InStock
29-0005-93
Affinity chromatography columns, His SpinTrap™ TALON®
Colonnes pour chromatographie
His SpinTrap™ TALON® is a single-use column designed for high purity, small-scale purification and screening of histidine-tagged proteins from clarified cell lysates.
  • Suitable for purification of multiple samples in parallel, such as screening
  • With a microcentrifuge, one run takes about 10 minutes
Convenient, ready-to-use single-use spin column, prepacked with 100 μl TALON® Superflow™ cobalt-IMAC (immobilized metal affinity chromatography) medium offering a different selectivity compared to nickel-charged media.
The columns make parallel preparations of few or many small-scale cell lysates quick and easy. Prepacked columns enable highly reproducible column to column purification and yield, with RSD <6%. Leakage of cobalt ions from TALON® Superflow™ medium is negligible.
His SpinTrap TALON® columns are used together with a standard microcentrifuge and purification takes approximately 10 minutes.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité