Filter plates, 96-well, His MultiTrap™ TALON®
Fournisseur: Cytiva
29-0005-96
29-0005-96EA 841 EUR InStock
29-0005-96
Filter plates, 96-well, His MultiTrap™ TALON®
Microplaques Plaques de filtration
His MultiTrap™ TALON® multiwell plates are designed for high-throughput screening of histidine-tagged proteins from clarified cell lysates. His MultiTrap™ TALON® prepacked plates are manufactured with high reproducibility with regards to the well and media volumes in order to enable consistent results. The average purity of His MultiTrap TALON® is >85% with yields up to 1 mg/well. Leakage of cobalt ions from TALON® Superflow™ medium is negligible. His MultiTrap™ TALON® is used manually with vacuum or centrifugation, or in a liquid handling station.
  • Up to 1 mg of pure histidine-tagged protein per well
  • Large sample volumes can be applied all at once
Prefilled with 50 µl TALON® Superflow™ cobalt-IMAC (immobilized metal affinity chromatography) medium for reproducible and high throughput parallel screening, offering a different selectivity compared to nickel-charged media.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité