Hydrophobic interaction chromatography columns, HiPrep™ Phenyl HP 16/10
Fournisseur: Cytiva
29-0181-84
29-0181-84EA 837 EUR InStock
29-0181-84
Hydrophobic interaction chromatography columns, HiPrep™ Phenyl HP 16/10
Colonnes pour chromatographie
HiPrep 16/10 Phenyl HP is a prepacked column designed for preparative hydrophobic interaction chromatography (HIC) with good resolution.
  • Aromatic HIC resin for intermediate purification and polishing
  • Selectivity similar to Phenyl Sepharose 6 Fast Flow (Low Sub) but higher resolution due to the smaller 34 µm bead size
  • Provides fast, easy high-capacity separations in a convenient format
Phenyl Sepharose HP chromatography resin is based on a matrix of highly cross-linked agarose beads, which have been derivatized with phenyl groups. These are attached to the agarose matrix via uncharged, chemically stable ether linkages. The result is a truly hydrophobic resin with no ionic properties.
Phenyl Sepharose HP chromatography resin is based on a matrix of highly cross-linked agarose beads, which have been derivatized with phenyl groups. These are attached to the agarose matrix via uncharged, chemically stable ether linkages. The result is a truly hydrophobic resin with no ionic properties.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité