Antibody column package, Modern Protein A (1-step purification protocol)
Fournisseur: Cytiva
29-4976-28
29-4976-28EA 259 EUR InStock
29-4976-28
Antibody column package, Modern Protein A (1-step purification protocol)
Colonnes pour chromatographie
The Antibody column package includes modern Protein A affinity resin and desalting columns for purification of human antibodies.
  • Contains columns for a 1-step protocol using affinity chromatography, HiTrap MabSelect PrismA 1×1 ml and HiTrap Desalting 1×5 ml
  • HiTrap MabSelect PrismA column is prepacked with resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant
  • HiTrap Desalting column is packed with Sephadex G-25 to give you quick pH neutralisation of the eluted antibodies
  • Includes instruction with guidance and tips to quickly get started
The package includes a HiTrap column prepacked with MabSelect PrismA resin containing a protein A-derived ligand produced in E. coli. Fermentation and subsequent purification are performed in the absence of animal products. The ligand has been specially engineered to create an affinity resin with enhanced alkali and protease stability. The specificity of binding to the Fc region of IgG is similar to that of conventional Protein A and provides excellent purification in one step.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité