Calcofluor white stain 5 mM dans l'eau colorant fluorescent
Fournisseur: Biotium
29067
29067.EA 96.6 EUR InStock
29067.
Calcofluor white stain 5 mM dans l'eau colorant fluorescent
Calcofluor white stain
Calcofluor White is a fluorescent blue dye that binds to cellulose and chitin, which can be found in the cell walls of fungi, algae, and plants. In budding yeast Calcofluor White stains bud scars more strongly, because they have a higher concentration of chitin. Calcofluor White is sometimes used to identify fungal infections in medical samples.
  • Yeast and fungus specific staining
  • Provided as a 5 mM solution in water
  • Ex/Em 360/430 nm
  • Store at -20°C, protected from light
Calcofluor White also stains gram-positive bacteria, but not gram-negative bacteria or mammalian cells.
Calcofluor White is also sold together with ViaVac Red/Green as part of the Yeast Vitality Staining Kit (31062)
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité