Synonyms:
Lithium tri-tert-butoxyaluminohydride
30716.09
30716.09EA
89.9
EUR
InStock
30716.09
Lithium tri-tert-butoxyaluminum hydride ≥94%
Lithium tri-tert-butoxyaluminum hydride
Purity: 94%
CAS No.: 17476-04-9
Molecular Formula: C12H28AlLiO3
Formula Weight: 254.28
Physical Form: Powder
Density: 1.03
Sensitivity: Moisture Sensitive
Appearance: White to pale grey
Odour: Odourless
Melting Point: ca 319° dec.
Beilstein Reference: 5796791
UN No.: UN1409
MDL No.: MFCD00011532