Supelco® TraceCERT® Organic Reference Standard, Flutolanil
Fournisseur: Merck
30988-50MG
30988-50MGEA 107 EUR InStock
30988-50MG
Supelco® TraceCERT® Organic Reference Standard, Flutolanil
Étalons Étalons organiques
The flutolanil CRM may also find following uses: Examine the reproductive endpoints on the gonad endocrine system of the parental zebrafish after chronic exposure to different concentrations of flutolanil for 60 days. Spectroscopic analysis of acetamiprid flutolanil and etofenprox residues extracted from tomato fruits. Development and validation of two robust chromatographic methods for estimation of tomatoes specific acetamiprid flutolanil and etofenprox residues on fruits before food processing and serving. To investigate the developmental toxicity and the potential mechanism of thyroid endocrine disruption induced by flutolanil in zebrafish embryos based on H¹ NMR metabolomics analysis. To develop a sensitive LC-MS/MS analytical method for the determination of residual flutolanil and its principal metabolites and conjugates in livestock and seafood products.
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Attention: P273 - P391 - P501
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité