32435.K2
32435.K2EA
58
EUR
InStock
32435.K2
Méthanol ≥99.8%, Ultrapure pour spectrophotométrie
Méthanol
Purity: 99.8+%
CAS No.: 67-56-1
Molecular Formula: CH3OH
Formula Weight: 32.04
Flash Point: 11°(52°F)
Physical Form: Liquid
Density: 0.791
Refractive Index: 1.3290
Odour: Mild
Melting Point: -98°
Boiling Point: 64.7°
Beilstein Reference: 1098229
Merck Reference: 14,5957
UN No.: 1230
MDL No.: MFCD00004595