Synonyms:
2,3,7,8-Tetrahydroxy[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione
M-Clarity™ quality level = MQ100
324683-500
324683-500EA
142
EUR
InStock
324683-500
Ellagic acid dihydrate ≥98% (par HPLC), Calbiochem®, Sigma-Aldrich®
Ellagic acid dihydrate
Ellagic Acid, Dihydrate, a cell-permeable, reversible, potent antioxidant that has anti-mutagenic and anti-carcinogenic properties.
Inhibits DNA topoisomerases I and II (IC50=1.8µM and 2.1µM, respectively). Also a potent inhibitor of PKA catalytic subunit (IC50=2µM) and of PKC (IC50=8µM).
Conditionnement: Packaged under inert gas