M-Clarity™ quality level = MQ100
324880-10
324880-10EA
121
EUR
InStock
324880-10
(-)-Epigallocatechin gallate ≥95% (par HPLC), Calbiochem®, Sigma-Aldrich®
(-)-Epigallocatechin gallate
One of the main polyphenolic constituents of green tea that exhibits potent antitumor, anti-inflammatory, and antioxidant properties
Arrests cell cycling at G0/G1 phases and induces apoptosis in a dose-dependent manner. EGCG has been shown to inhibit PMA-induced skin thickening, and the activation of protein kinase C. Also inhibits the activation of ornithine decarboxylase, and activation of interleukin-1[alpha] mRNA and protein expression. Inhibits inducible nitric oxide synthase (iNOS) gene expression and enzyme activity and inhibits peroxynitrite-mediated formation of 8-oxodeoxyguanosine and 3-nitrotyrosine. Also strongly and directly inhibits telomerase.
Conditionnement: Packaged under inert gas