34673-2.5L
34673-4X4L
34673-2.5LEA
161
EUR
InStock
34673-2.5L
34673-4X4L
Acide formique, CHROMASOLV™ LC-MS LabReady™, Mélange de solvants pour LC-MS, Riedel-de Haën™
Acide formique
- For LC-MS and LC-UV trace metals analysis
- HPLC / UV-VIS quality control
- Low gradient baseline drift
- Low levels of trace metals
- No non-volatile impurities
RTECS Number: 231-791-2