35480.06
35480.06EA
142
EUR
InStock
35480.06
Oxyde de béryllium ≥99.95% (base métaux)
Oxyde de béryllium
Purity: 99.95%
CAS No.: 1304-56-9
Molecular Formula: BeO
Formula Weight: 25.01
Physical Form: Powder
Density: 3.01
Melting Point: 2575°
Boiling Point: 4300°
Merck Reference: 14,1172
UN No.: 1566
MDL No.: MFCD00003457