35592.K2
35592.K2EA
34.7
EUR
InStock
35592.K2
Potassium hydroxyde solution standardisée 1.0 N en solution aqueuse
Potassium hydroxyde
CAS No.: 1310-58-3
Molecular Formula: KOH
Formula Weight: 56.11
Physical Form: Liquid
Appearance: Colourless
UN No.: 1814
MDL No.: MFCD00003553