35607.K2
35607.K2EA
68
EUR
InStock
35607.K2
Acide chlorhydrique 50% (v/v) en solution aqueuse
Acide chlorhydrique
CAS No.: 7647-01-0
Molecular Formula: HCl
Formula Weight: 36.46
Physical Form: Liquid
Appearance: Colourless to pale yellow
UN No.: 1789
MDL No.: MFCD00011324