35610.K2
35610.K2EA
57.5
EUR
InStock
35610.K2
Acide sulfurique solution standardisée 6.0 N en solution aqueuse
Acide sulfurique
CAS No.: 7664-93-9
Molecular Formula: H2SO4
Formula Weight: 98.08
Physical Form: Liquid
Appearance: Colourless to pale yellow
Odour: Acrid
UN No.: 2796
MDL No.: MFCD00064589