35631.K2
35631.K2EA
37.9
EUR
InStock
35631.K2
Sodium hydroxyde solution standardisée 2.0 N en solution aqueuse
Sodium hydroxyde
CAS No.: 1310-73-2
Molecular Formula: NaOH
Formula Weight: 40.00
Physical Form: Liquid
Odour: Odourless
UN No.: 1824
MDL No.: MFCD00003548