Dimethylpolysiloxane set of analytical standards, molecular weight series
Fournisseur: Merck
361879-1SET
361879-1SETEA 364 EUR InStock
361879-1SET
Dimethylpolysiloxane set of analytical standards, molecular weight series
Étalons Étalons de chromatographie gazeuse
Clear, colorless fluid polymer useful as a stationary phase in gas chromatography and as an anti-foaming agent.
It was used in a study performed to understand its molecular weight and molecular weight distribution by off-line coupling of GPC with MALDI mass spectrometer.
Kit components
Mw 40000
Mw 100000
Mw 270000
Certifications: Exact values can be taken from the accompanying certificate of analysis.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f21ae9001b503855ae4dcd70d240836869bf2bb7&vid=f21ae9001b503855ae4dcd70d240836869bf2bb7&prid=web&clid=SAPDAM
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité