Synonyms:
Lutetium oxide, Dilutetium trioxide
36244.03
36244.03EA
209
EUR
InStock
36244.03
Lutétium (III) oxyde ≥99.9% (REO, base d'oxyde de terre rare)
Lutétium (III) oxyde
Purity: 99.9%
CAS No.: 12032-20-1
Molecular Formula: Lu2O3
Formula Weight: 397.94
Physical Form: 3-12mm Sintered Lump
Density: 9.420
Melting Point: 2487°
Merck Reference: 14,5615
MDL No.: MFCD00011100