Synonyms:
Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane
Stabilization:
Stabilised with ethanol 0.5-1.0%
366927-2.5L
366927-2.5LEA
166
EUR
InStock
366927-2.5L
Chloroforme ≥99.8% stabilisé, CHROMASOLV™ pour HPLC, Riedel-de Haën™
Chloroforme
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Beilstein Number: 1731042