374776-5G
374776-5GEA
76.8
EUR
InStock
374776-5G
4-Méthoxybiphényle 97%, Sigma-Aldrich®
4-Méthoxybiphényle
4-Methoxybiphenyl has been used as a standard reagent whose fluorescence intensity is associated with the fluorescence characteristics of the products of derivatisation reaction for aryl halides with phenylboronic acid (PBA).