38595.14
38595.14EA
711
EUR
InStock
38595.14
Lithium hexafluoroantimonate ≥97%
Lithium hexafluoroantimonate
Purity: 97%
CAS No.: 18424-17-4
Molecular Formula: LiSbF6
Formula Weight: 242.68
Physical Form: Powder
Sensitivity: Hygroscopic
Boiling Point: dec.
Solubility: Very soluble in water
UN No.: 1549
MDL No.: MFCD00042319