VWR®, Bottles, PET, Square, with Screw Cap
Fournisseur: VWR Collection
391-0629EA 84.5 EUR InStock
391-0629 391-0628 391-0627 391-0631 391-0630 391-0626 391-0648
VWR®, Bottles, PET, Square, with Screw Cap
Flacons Flacons en plastique
Square PET media bottles that are designed for packaging, storing, or transporting powder and liquid samples.
  • Square design allows efficient use of storage space
  • PET resin is lightweight, break-resistant and transparent
  • Bottles feature moulded-in graduations
  • EB Sterilisation Sterility Assurance Level (SAL) of 10⁻⁶ (ISO 11137)
  • Non pyrogenic
VWR® square PET media bottles are manufactured in a Class 100 000 cleanroom environment. Manufactured under EN ISO 13485:2016.
Certifications: Bottles meet the requirements of BSE/TSE, the BPA statement and heavy metals ROHS.
Conditionnement: Bottles are available individually wrapped, or in tray packs (triple-bagged PET bottles for cleanroom applications).
Attention: These bottles are not to be used for production packaging.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité