Synonyms:
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
Packaged under argon.
39289.ME
39289.MEEA
313
EUR
InStock
39289.ME
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
Bis(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulphonate
CAS No.: 99326-34-8
Molecular Formula: C17H24F3O3RhS
Formula Weight: 468.34
Physical Form: Crystalline, Packaged under argon
Sensitivity: Air Sensitive
Solubility: Soluble in chloroform
MDL No.: MFCD00143752