Synonyms:
Zirconate(2-), hexafluoro-, Hexafluorozirconic acid, Hexafluorozirconate(2-)
39462.A3
39462.A3EA
208
EUR
InStock
39462.A3
Dihydrogen hexafluorozirconate 45% (p/p) en solution aqueuse
Dihydrogen hexafluorozirconate
CAS No.: 12021-95-3
Molecular Formula: H2ZrF6
Formula Weight: 207.22
Physical Form: Liquid
Density: 1.51
Appearance: Colourless
UN No.: UN2922
MDL No.: MFCD00082965