39563.14
39563.14EA
1100
EUR
InStock
39563.14
Dimethylzinc ≥96%
Dimethylzinc
Purity: 96%
CAS No.: 544-97-8
Molecular Formula: (CH3)2Zn
Formula Weight: 95.44
Flash Point: -18°(-0.4°F)
Physical Form: Liquid, packaged in a non-returnable cylinder
Density: 1.386
Sensitivity: Air and Moisture Sensitive
Melting Point: -40°
Boiling Point: 46°
Beilstein Reference: 3587195
Merck Reference: 14,3263
UN No.: 3394
MDL No.: MFCD00014854