Packaged under argon.
39996.04
39996.04EA
465
EUR
InStock
39996.04
Scandium ≥99.9% (REO, base d'oxyde de terre rare), pièces, dendritique distillée
Scandium
Purity: 99.9%
CAS No.: 7440-20-2
Physical Form: Packaged under argon
Appearance: Grey
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00016323