Dichlorhydrate de Diamidino-4',6-phénylindole-2 (DAPI) utilisé en coloration nucléaire
Fournisseur: Biotium
40011
40011.EA 174 EUR InStock
40011.
Dichlorhydrate de Diamidino-4',6-phénylindole-2 (DAPI) utilisé en coloration nucléaire
Dichlorhydrate de Diamidino-4',6-phénylindole-2 (DAPI)
DAPI (4′,6-Diamidino-2-Phenylindole, dihydrochloride) is a popular blue DNA dye that is used as a nuclear counterstain in fluorescence microscopy, chromosome staining and flow cytometry. The dye binds to the minor groove of dsDNA with approximately 20-fold fluorescence enhancement, with higher affinity for A-T rich regions. Also see DAPI dilactate (catalog no. 40009), a more water soluble form of the dye.
  • Classic blue nuclear counterstain for fixed cells
  • Can be used at higher concentrations to stain live cells
  • λEx/λEm (with DNA) = 358/461 nm
  • Off-white solid soluble in H2O
  • Store at 4°C and protect from light, especially in solution
DAPI can be used to stain live bacteria (gram-positive and gram-negative) but in live yeast the staining is weak and not nuclear.
Formule: C₁₆H₁₅N₅·2HCl
Température de stockage: Réfrigérateur
Numéro MDL: MFCD00012681
Numéro CAS: 28718-90-3
Commander maintenant
COMMANDE
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité