Synonyms:
Argentous iodate
40109.14
40109.14EA
416
EUR
InStock
40109.14
Iodate d'argent 98%
Iodate d'argent
Purity: 99%
CAS No.: 7783-97-3
Molecular Formula: AgIO3
Formula Weight: 282.77
Physical Form: Powder
Density: 5.53
Appearance: White
Odour: Odourless
Melting Point: 200°
Solubility: Practically insoluble in water. Soluble in aqueous ammonia
Merck Reference: 14,8515
UN No.: 1479
MDL No.: MFCD00014150