40114.36
40114.36EA
41.8
EUR
InStock
40114.36
di-Sodium tétraborate décahydraté 99.5-105.0% ACS
di-Sodium tétraborate décahydraté
Purity: 99.5-105.0%
CAS No.: 1303-96-4
Molecular Formula: B4Na2O7.10H2O
Formula Weight: 381.37 (201.22anhy)
Physical Form: Powder
Density: 1.73
Appearance: Grey, blue, or green
Odour: Odourless
pH: 9,24
Melting Point: 75°
Solubility: Soluble in water, glycerol. Insoluble in alcohol
Merck Reference: 14,8590
MDL No.: MFCD00149193