40120.36
40120.36EA
116
EUR
InStock
40120.36
Hydrazinium sulfate ≥99.0% ACS
Hydrazinium sulfate
Purity: 99.0% min
CAS No.: 10034-93-2
Molecular Formula: H2NNH2.H2SO4
Formula Weight: 130.12
Physical Form: Crystalline
Density: 1.370
pH: 1.3
Melting Point: ca 255° dec.
Merck Reference: 14,4772
UN No.: 3288
MDL No.: MFCD00044873
Reagent for determination of C-terminal amino acids