40230.14
40230.14EA
200
EUR
InStock
40230.14
Dichromate de césium ≥99.8% (base métaux)
Dichromate de césium
Purity: 99.8%
CAS No.: 13530-67-1
Molecular Formula: Cr2Cs2O7
Formula Weight: 481.80
Physical Form: Coarse Powder
Sensitivity: Hygroscopic
UN No.: 3087
MDL No.: MFCD00049436