Supelco® TraceCERT® Organic Reference Standard, Fenpropimorph
Fournisseur: Merck
40713-100MG
40713-100MGEA 125 EUR InStock
40713-100MG
Supelco® TraceCERT® Organic Reference Standard, Fenpropimorph
Étalons Étalons organiques
Fenpropimorph is a systematic fungicide that belongs to the morpholine group of sterol biosynthesis inhibitors. It is widely applied in cereals to control pathogens such as Erysiphe graminis (powdery mildew) and Puccinia recondita (cereal rust). Fenpropimorph inhibits fungal; 8 to 7 isomerase and 14 reductase enzymes involved in the ergosterol biosynthesis. Fenpropimorph is not approved for use in European Union (EU) as per Regulation (EC) No 1107/2009. The maximum residue level (MRLs) for fenpropimorph (sum of isomers) following Reg. (EU) 2019/552 has been set from 0,01 to 3 mg/kg for products of plant and animal origin. Fenpropimorph has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP) run within the EU and EFTA in/on products of plant origin.
Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Attention: P201 - P202 - P273 - P301 + P312 - P302 + P352 - P308 + P313
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité