40870.KS
40870.KSEA
245
EUR
InStock
40870.KS
Nickel chromium alloy (80:20 w%) ≥99.95% (base métaux), cible de pulvérisation, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Nickel chromium alloy (80:20 w%)
Purity: 99.95%
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Odour: Odourless
MDL No.: MFCD00673110
Magnetron sputtering source