40874.KS
40874.KSEA
634
EUR
InStock
40874.KS
Nickel ≥99.99% (base métaux), cible de pulvérisation, Ø 50.8 mm (2.0 in), Thickness: 3.18 mm (0.125 in)
Nickel
Purity: 99.99%
CAS No.: 7440-02-0
Odour: Odourless
MDL No.: MFCD00011137
Magnetron sputtering source