Synonyms:
Gadolinium(III) i-propoxide
41377.03
41377.03EA
255
EUR
InStock
41377.03
Gadolinium(III) isopropoxide ≥99% (base métaux)
Gadolinium(III) isopropoxide
Purity: 99%
CAS No.: 14532-05-9
Molecular Formula: C9H21GdO3
Formula Weight: 334.51
Physical Form: Powder
Sensitivity: Air and Moisture Sensitive
Appearance: Off-white
Solubility: Soluble in anhydrous 2-propanol, toluene
UN No.: UN2925
MDL No.: MFCD00144381
Applications: Sol-Gel synthesis of gadolinium compounds