41558.FF
41558.FFEA
348
EUR
InStock
41558.FF
Lanthane ≥99.9% (REO, base d'oxyde de terre rare), en feuilles, Thickness: 0.1 mm (0.004 in)
Lanthane
Purity: 99.9%
CAS No.: 7439-91-0
Physical Form: ≈0.39g/25x25mm, Packaged in mylar under argon
Appearance: Gray to silver
UN No.: 3208
MDL No.: MFCD00011066