41771.36
41771.36EA
66.6
EUR
InStock
41771.36
Anhydride phtalique 99.0-100.2% ACS
Anhydride phtalique
Purity: 99.0-100.2%
CAS No.: 85-44-9
Molecular Formula: C6H4(CO)2O
Formula Weight: 148.12
Flash Point: 152°(305°F)
Physical Form: Flaky Crystals
Density: 1.53
Sensitivity: Moisture Sensitive
Odour: Weak, characteristic
Melting Point: 131-133°
Boiling Point: 295° subl.
Beilstein Reference: 118515
Merck Reference: 14,7372
UN No.: 2214
MDL No.: MFCD00005918