Packaged under argon in resealable ChemSeal™ bottles.
41839.AK
41839.AKEA
39.7
EUR
InStock
41839.AK
Tert-Butyle methyle oxyde 99+% pour HPLC
Méthoxy-2-méthyl-2 propane
Purity: 99+%
CAS No.: 1634-04-4
Molecular Formula: (CH3)3COCH3
Formula Weight: 88.15
Flash Point: -25°(-13°F)
Physical Form: Liquid
Density: 0.742
Refractive Index: 1.3690
Odour: Mild
Melting Point: -109°
Boiling Point: 55-56°
Beilstein Reference: 1730942
Merck Reference: 14,6032
UN No.: 2350
MDL No.: MFCD00008812
Chromatography solvent