42094.03
42094.03EA
144
EUR
InStock
42094.03
Triiodure de thulium ≥99.9% (REO, base d'oxyde de terre rare)
Triiodure de thulium
Purity: 99.9%
CAS No.: 13813-43-9
Molecular Formula: TmI3
Formula Weight: 549.65
Physical Form: -20 Mesh
Melting Point: 1015°
Boiling Point: 1260°
MDL No.: MFCD00050141