42096.03
42096.03EA
200
EUR
InStock
42096.03
Chloropentacarbonylrhénium ≥98%
Chloropentacarbonylrhénium
Purity: 98%
CAS No.: 14099-01-5
Molecular Formula: C5ClO5Re
Formula Weight: 361.71
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: White to pale brown
UN No.: 3466
MDL No.: MFCD00013296