42105-1L
42105-1LEA
570
EUR
InStock
42105-1L
Méthanol ≥99.9%, TraceSELECT™ pour analyse de la spéciation des métaux, Riedel-de Haën™
Méthanol
High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.