42330.QK
42330.QKEA
276
EUR
InStock
42330.QK
Nickel ≥99.995% (base métaux), cartouche, Puratronic®, Length: 12.7 mm (0.50 in), Ø 6.35 mm (0.25 in)
Nickel
Purity: 99.995%
CAS No.: 7440-02-0
Physical Form: ≈3.58g/slug
Odour: Odourless
MDL No.: MFCD00011137