42331.QK
42331.QKEA
292
EUR
InStock
42331.QK
Nickel ≥99.995% (base métaux), cartouche, Puratronic®, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Nickel
Purity: 99.995%
CAS No.: 7440-02-0
Physical Form: ≈1.8g/slug
Odour: Odourless
MDL No.: MFCD00011137