42366.18
42366.18EA
167
EUR
InStock
42366.18
Nickel chromium alloy (80:20 w%) ≥99.95% (base métaux), cartouche, Length: 6.35 mm (0.25 in), Ø 6.35 mm (0.25 in)
Nickel chromium alloy (80:20 w%)
Purity: 99.95%
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈1.7g/slug
Odour: Odourless