42368.18
42368.18EA
283
EUR
InStock
42368.18
Nickel chromium alloy (80:20 w%) ≥99.95% (base métaux), cartouche, Length: 3.175 mm (0.125 in), Ø 3.175 mm (0.125 in)
Nickel chromium alloy (80:20 w%)
Purity: 99.95%
CAS No.: 11106-97-1
Molecular Formula: Ni:Cr; 80:20 wt%
Physical Form: ≈0.22g/slug
Odour: Odourless