42552.M4
42552.M4EA
368
EUR
InStock
42552.M4
Acide sulfurique 10% (v/v) en solution aqueuse
Acide sulfurique
CAS No.: 7664-93-9
Molecular Formula: H2O4S
Formula Weight: 98.08
Physical Form: Liquid
Appearance: Colourless
UN No.: 2796
MDL No.: MFCD00064589